Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyre glacial ≥99%

Eddikesyre glacial ≥99%

Glass bottle, 1 L
Catalog # 33252.K2
Corrosive
Flammable
Eddikesyre glacial ≥99%
Catalog # 33252.K2
CAS Number:  64-19-7
Eddikesyre glacial ≥99%
Eddikesyre glacial ≥99%

Glass bottle, 1 L

CAS Number:  64-19-7
Price
864,00 kr
Log in for availability
1 * 1 L
Product Details & Documents

Purity: 99+% CAS No.: 64-19-7 Molecular Formula: CH3CO2H Formula Weight: 60.05 Flash Point: 40°(104°F) Physical Form: Liquid Density: 1.049 Refractive Index: 1.3721 Odour: Pungent pH: 2,5 Melting Point: 16.6° Boiling Point: 118.1° Merck Reference: 14,55 UN No.: 2789 MDL No.: MFCD00036152

Synonyms:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS Number:64-19-7
  • Formula:H₃CCOOH
  • MDL Number:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Merck Index:14,55
  • Boiling Pt:118,1 °C
  • Melting Pt:16,6 °C
  • Density:1,049
  • Flash Pt:40 °C (104 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:200-580-7
  • UN:2789
  • ADR:8,II
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.