Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyre glacial ≥99%

Eddikesyre glacial ≥99%

Glass bottle, 250 mL
Katalog # 33252.AK
Ætsende
Brandfarlig
Leverandør:   Thermo Fisher Scientific
Eddikesyre glacial ≥99%
Katalog # 33252.AK
CAS nummer:  64-19-7
Eddikesyre glacial ≥99%
Eddikesyre glacial ≥99%

Glass bottle, 250 mL

CAS nummer:  64-19-7
Price
545,00 kr
Log ind for tilgængelighed
1 * 250 mL
Product Details & Documents

Purity: 99+% CAS No.: 64-19-7 Molecular Formula: CH3CO2H Formula Weight: 60.05 Flash Point: 40°(104°F) Physical Form: Liquid Density: 1.049 Refractive Index: 1.3721 Odour: Pungent pH: 2,5 Melting Point: 16.6° Boiling Point: 118.1° Merck Reference: 14,55 UN No.: 2789 MDL No.: MFCD00036152

Synonymer:  Vinegar acid, Methanecarboxylic acid, Ethanoic acid
  • CAS nummer:64-19-7
  • Formel:H₃CCOOH
  • MDL nummer:MFCD00036152
  • Molecular Weight:60.05 g/mol
  • Merck Index:14,55
  • Kogende Pt:118,1 °C
  • Smeltende Pt:16,6 °C
  • Tæthed:1,049
  • Flash Pt:40 °C (104 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-580-7
  • FN:2789
  • ADR:8,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    250 mL

Specifikationer