Orders LinkContactUsLinkComponent
Rods, ceramic, round, solid, AL23
Rods, ceramic, round, solid, AL23
0,6 mm, 100 mm
Katalog # 32921.DM
Leverandør:   Thermo Fisher Scientific
Rods, ceramic, round, solid, AL23
Katalog # 32921.DM
Rods, ceramic, round, solid, AL23
0,6 mm, 100 mm
Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 mm
Product Details & Documents
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.
Specifikationer
  • Udv.Ø:
    0,6 mm
  • Længde:
    100 mm
  • Seq:
    05