Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bromphenol blå ACS

Bromphenol blå ACS

Glass bottle for solids, 25 g
Katalog # 32641.14
Leverandør:   Thermo Fisher Scientific
Bromphenol blå ACS
Katalog # 32641.14
CAS nummer:  115-39-9
Bromphenol blå ACS
Bromphenol blå ACS

Glass bottle for solids, 25 g

CAS nummer:  115-39-9
Price
1.480,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

CAS No.: 115-39-9 Molecular Formula: C19H10Br4O5S Formula Weight: 669.98 Physical Form: Crystalline Odour: Odourless Melting Point: 270° dec. Beilstein Reference: 61698 Merck Reference: 14,1444 MDL No.: MFCD00005875

Synonymer:  Bromphenol Blue Sultone Form, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein
  • CAS nummer:115-39-9
  • Formel:C₁₉H₁₀Br₄O₅S
  • MDL nummer:MFCD00005875
  • Molecular Weight:669.97 g/mol
  • Merck Index:14,1444
  • Smeltende Pt:270 °C (dec.)
  • Tæthed:1 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-086-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer