Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Hexan ≥95% for spectrophotometry

n-Hexan ≥95% for spectrophotometry

4x1 L, 4
Catalog # 32454.M1
Flammable
Health
Irritant
Environment
n-Hexan ≥95% for spectrophotometry
Catalog # 32454.M1
CAS Number:  110-54-3
n-Hexan ≥95% for spectrophotometry
n-Hexan ≥95% for spectrophotometry

4x1 L, 4

CAS Number:  110-54-3
Price
3.810,00 kr
Log in for availability
1 * 4 Items
Product Details & Documents

Purity: 95+% CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520

Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:14,4694
  • Boiling Pt:69 °C
  • Melting Pt:–95 °C
  • Density:0,659
  • Flash Pt:–23 °C (–9 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
Specifications

Specifications

  • Pakn:
    4
  • Pk Info:
    4x1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.