Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Hexan ≥95% for spectrophotometry

n-Hexan ≥95% for spectrophotometry

Glass bottle, 4 L
Katalog # 32454.K7
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
n-Hexan ≥95% for spectrophotometry
Katalog # 32454.K7
CAS nummer:  110-54-3
n-Hexan ≥95% for spectrophotometry
n-Hexan ≥95% for spectrophotometry

Glass bottle, 4 L

CAS nummer:  110-54-3
Price
3.400,00 kr
Log ind for tilgængelighed
1 * 4 L
Product Details & Documents

Purity: 95+% CAS No.: 110-54-3 Molecular Formula: CH3(CH2)4CH3 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Odour: Weak, characteristic Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 Merck Reference: 14,4694 UN No.: 1208 MDL No.: MFCD00009520

Synonymer:  Hexyl hydride, Hexanen, Hexane
  • CAS nummer:110-54-3
  • Formel:H₃C(CH₂)₄CH₃
  • MDL nummer:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:14,4694
  • Kogende Pt:69 °C
  • Smeltende Pt:–95 °C
  • Tæthed:0,659
  • Flash Pt:–23 °C (–9 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-777-6
  • FN:1208
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    4 L

Specifikationer