Isooctan 99+% ACS
4x1 L, 4
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99+% CAS No.: 540-84-1 Molecular Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 Flash Point: -7°(19°F) Physical Form: Liquid Density: 0.692 Refractive Index: 1.3910 Odour: Nearly Odourless Melting Point: -107° Boiling Point: 98-99° Solubility: Soluble in benzene, toluene, xylene, chloroform, ether, CS2, CCl4, and DMF Beilstein Reference: 1696876 Merck Reference: 14,5193 UN No.: 1262 MDL No.: MFCD00008943
Solvent, thinner, in determination of fuel octane numbers
Synonymer: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
- CAS nummer:540-84-1
- Formel:(CH₃)₂CHCH₂C(CH₃)₃
- MDL nummer:MFCD00008943
- Molecular Weight:114.23 g/mol
- Merck Index:14,5193
- Kogende Pt:98…99 °C
- Smeltende Pt:–107 °C
- Tæthed:0,692
- Flash Pt:–7 °C (19 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-759-1
- FN:1262
- ADR:3,II
Documents
Specifikationer
Specifikationer
- Pakn:4
- Pk Info:4x1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



