Isooctan 99+% ACS
Glass bottle, 1 L
Purity: 99+% CAS No.: 540-84-1 Molecular Formula: (CH3)2CHCH2C(CH3)3 Formula Weight: 114.23 Flash Point: -7°(19°F) Physical Form: Liquid Density: 0.692 Refractive Index: 1.3910 Odour: Nearly Odourless Melting Point: -107° Boiling Point: 98-99° Solubility: Soluble in benzene, toluene, xylene, chloroform, ether, CS2, CCl4, and DMF Beilstein Reference: 1696876 Merck Reference: 14,5193 UN No.: 1262 MDL No.: MFCD00008943
Solvent, thinner, in determination of fuel octane numbers
: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
- :540-84-1
- :(CH₃)₂CHCH₂C(CH₃)₃
- :MFCD00008943
- :114.23 g/mol
- :14,5193
- :98…99 °C
- :–107 °C
- :0,692
- :–7 °C (19 °F)
- :Stue temperatur
- :208-759-1
- :1262
- :3,II
- Pakning:Glass bottle
- Pakn:1 L



