Orders LinkContactUsLinkComponent
Methylethylketon ≥97%
4x1 L, 4
  31069.M1
Methylethylketon ≥97%
  31069.M1
 :  78-93-3
Methylethylketon ≥97%
Methylethylketon ≥97%
4x1 L, 4
 :  78-93-3
2.260,00 kr
1 * 4 Items
Purity: 97% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.805 Refractive Index: 1.3790 Appearance: Colourless Odour: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648
 :  Ethylmethylketon, Butanon
  •  :78-93-3
  •  :C₂H₅COCH₃
  •  :MFCD00011648
  •  :72.11 g/mol
  •  :14,6072
  •  :80 °C
  •  :–87 °C
  •  :0,805
  •  :–3 °C (26 °F)
  •  :Stue temperatur
  •  :201-159-0
  •  :1193
  •  :3,II
  •  :01-2119457290-43
  • Pakn:
    4
  • Pk Info:
    4x1 L