Methylethylketon ≥97%
4x1 L, 4
Purity: 97% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.805 Refractive Index: 1.3790 Appearance: Colourless Odour: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648
: Ethylmethylketon, Butanon
- :78-93-3
- :C₂H₅COCH₃
- :MFCD00011648
- :72.11 g/mol
- :14,6072
- :80 °C
- :–87 °C
- :0,805
- :–3 °C (26 °F)
- :Stue temperatur
- :201-159-0
- :1193
- :3,II
- :01-2119457290-43
- Pakn:4
- Pk Info:4x1 L