Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylethylketon ≥97%

Methylethylketon ≥97%

Glass bottle, 1 L
Katalog # 31069.K2
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Methylethylketon ≥97%
Katalog # 31069.K2
CAS nummer:  78-93-3
Methylethylketon ≥97%
Methylethylketon ≥97%

Glass bottle, 1 L

CAS nummer:  78-93-3
Price
698,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Purity: 97% CAS No.: 78-93-3 Molecular Formula: C4H8O Formula Weight: 72.11 Flash Point: -3°(26°F) Physical Form: Liquid Density: 0.805 Refractive Index: 1.3790 Appearance: Colourless Odour: Characteristic Melting Point: -87° Boiling Point: 80° Beilstein Reference: 741880 Merck Reference: 14,6072 UN No.: 1193 MDL No.: MFCD00011648

Synonymer:  Ethylmethylketon, Butanon
  • CAS nummer:78-93-3
  • Formel:C₂H₅COCH₃
  • MDL nummer:MFCD00011648
  • Molecular Weight:72.11 g/mol
  • Merck Index:14,6072
  • Kogende Pt:80 °C
  • Smeltende Pt:–87 °C
  • Tæthed:0,805
  • Flash Pt:–3 °C (26 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:201-159-0
  • FN:1193
  • ADR:3,II
  • NÅ:01-2119457290-43
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.