Orders LinkContactUsLinkComponent
Zinc ferrite
Zinc ferrite
100 g
Katalog # 19407.22
Leverandør:   Thermo Fisher Scientific
Zinc ferrite
Katalog # 19407.22
CAS nummer:  12063-19-3
Zinc ferrite
Zinc ferrite
100 g
CAS nummer:  12063-19-3
Price
4.380,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
CAS No.: 12063-19-3 Molecular Formula: ZnFe2O4 Formula Weight: 241.06 Physical Form: Fine Powder Odour: Odourless MDL No.: MFCD00049621
  • CAS nummer:12063-19-3
  • Formel:Fe₂H₂O₄Zn
  • MDL nummer:MFCD00049621
  • Molecular Weight:243.082 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:235-052-5
Specifikationer
Specifikationer
  • Pakn:
    100 g