Zinc ferrite
25 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
CAS No.: 12063-19-3 Molecular Formula: ZnFe2O4 Formula Weight: 241.06 Physical Form: Fine Powder Odour: Odourless MDL No.: MFCD00049621
- CAS nummer:12063-19-3
- Formel:Fe₂H₂O₄Zn
- MDL nummer:MFCD00049621
- Molecular Weight:243.082 g/mol
- Opbevaringstemperatur:Stue temperatur
- EINECS:235-052-5
Documents
Specifikationer
Specifikationer
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



