Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Propanol ≥99.5% semiconductor grade

2-Propanol ≥99.5% semiconductor grade

Glass bottle, 1 L
Katalog # 19397.K2
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
2-Propanol ≥99.5% semiconductor grade
Katalog # 19397.K2
CAS nummer:  67-63-0
2-Propanol ≥99.5% semiconductor grade
2-Propanol ≥99.5% semiconductor grade

Glass bottle, 1 L

CAS nummer:  67-63-0
Price
848,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents

Purity: 99.5% min CAS No.: 67-63-0 Molecular Formula: (CH3)2CHOH Formula Weight: 60.10 Flash Point: 12°(53°F) Physical Form: Liquid Density: 0.785 Refractive Index: 1.3770 Odour: Alcohol-like Melting Point: -90° Boiling Point: 82-83° Beilstein Reference: 635639 Merck Reference: 14,5208 UN No.: 1219 MDL No.: MFCD00011674

Synonymer:  Isopropylalkohol
  • CAS nummer:67-63-0
  • Formel:(CH₃)₂CHOH
  • MDL nummer:MFCD00011674
  • Molecular Weight:60.1 g/mol
  • Merck Index:14,5208
  • Kogende Pt:82…83 °C
  • Smeltende Pt:–90 °C
  • Tæthed:0,785
  • Flash Pt:12 °C (53 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-661-7
  • FN:1219
  • ADR:3,II
  • NÅ:01-2119457558-25
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.