Styren ≥99.5% stabilised
Plastic bottle, 1 L
Stabilized with 4-tert-butylcatechol
Purity: 99.5% CAS No.: 100-42-5 Molecular Formula: C8H8 Formula Weight: 104.15 Storage Temperature: 2°C to 8°C Flash Point: 31°(88°F) Physical Form: Liquid Density: 0.906 Refractive Index: 1.5460 Sensitivity: Light Sensitive Appearance: Colourless Odour: Pungent pH: neutral Melting Point: -31° Boiling Point: 145-146° Beilstein Reference: 1071236 Merck Reference: 14,8860 UN No.: 2055 MDL No.: MFCD00008612
: Phenylethene, Phenylethylene, Vinylbenzene
- :100-42-5
- :C₈H₈
- :MFCD00008612
- :104.15 g/mol
- :14,8860
- :145…146 °C
- :–31 °C
- :0,906
- :31 °C (88 °F)
- :Køleskab
- :2055
- :3,III
- :stabiliseret med 4-tert-Butylpyrocatechol
- :202-851-5
Documents
- Pakning:Plastic bottle
- Pakn:1 L