Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumazid ≥99%

Glass bottle for solids, 500 g
Katalog # 14314.36
Giftig
Miljø
Leverandør:   Thermo Fisher Scientific
Natriumazid ≥99%
Katalog # 14314.36
CAS nummer:  26628-22-8
Natriumazid ≥99%
Natriumazid ≥99%

Glass bottle for solids, 500 g

CAS nummer:  26628-22-8
Price
1.820,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 26628-22-8 Molecular Formula: N3Na Formula Weight: 65.01 Physical Form: Powder or Granular Density: 1.846 Appearance: White Odour: Odourless pH: 10 Melting Point: ca 275° dec Solubility: Soluble in water and liquid ammonia. Slightly soluble in alcohol Merck Reference: 14,8581 UN No.: 1687 MDL No.: MFCD00003536

In airbag inflation, as a preservative in diagnostic medicinals

Synonymer:  Sodium triazide
  • CAS nummer:26628-22-8
  • Formel:NaN₃
  • MDL nummer:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Merck Index:14,8581
  • Smeltende Pt:∼ 275 °C (dec.)
  • Tæthed:1,846
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:247-852-1
  • FN:1687
  • ADR:6.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer