Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lithiumbromid, anhydrous ≥99%

Lithiumbromid, anhydrous ≥99%

Plastic bottle for solids, 500 g
Katalog # 13408.36
Irriterende
Leverandør:   Thermo Fisher Scientific
Lithiumbromid, anhydrous ≥99%
Katalog # 13408.36
CAS nummer:  7550-35-8
Lithiumbromid, anhydrous ≥99%
Lithiumbromid, anhydrous ≥99%

Plastic bottle for solids, 500 g

CAS nummer:  7550-35-8
Price
1.640,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% min CAS No.: 7550-35-8 Molecular Formula: BrLi Formula Weight: 86.85 Physical Form: Crystalline Density: 3.464 Appearance: White Odour: Odourless Melting Point: 550° Boiling Point: 1265° Solubility: Very soluble in water and alcohol. Soluble in methanol, acetone, and glycol. Slightly soluble in pyridine Merck Reference: 14,5526 MDL No.: MFCD00011077

Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium

  • CAS nummer:7550-35-8
  • Formel:LiBr
  • MDL nummer:MFCD00011077
  • Molecular Weight:86.84 g/mol
  • Merck Index:14,5526
  • Kogende Pt:1265 °C
  • Smeltende Pt:550 °C
  • Tæthed:3,464
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-439-8
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    500 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.