Orders LinkContactUsLinkComponent
Lithiumbromid, anhydrous ≥99%
Glass bottle for solids, 100 g
  13408.22
Lithiumbromid, anhydrous ≥99%
  13408.22
 :  7550-35-8
Lithiumbromid, anhydrous ≥99%
Lithiumbromid, anhydrous ≥99%
Glass bottle for solids, 100 g
 :  7550-35-8
640,00 kr
1 * 100 g
Purity: 99% min CAS No.: 7550-35-8 Molecular Formula: BrLi Formula Weight: 86.85 Physical Form: Crystalline Density: 3.464 Appearance: White Odour: Odourless Melting Point: 550° Boiling Point: 1265° Solubility: Very soluble in water and alcohol. Soluble in methanol, acetone, and glycol. Slightly soluble in pyridine Merck Reference: 14,5526 MDL No.: MFCD00011077
Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium
  •  :7550-35-8
  •  :LiBr
  •  :MFCD00011077
  •  :86.84 g/mol
  •  :14,5526
  •  :1265 °C
  •  :550 °C
  •  :3,464
  •  :Stue temperatur
  •  :231-439-8
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g