Lithiumbromid, anhydrous ≥99%
Glass bottle for solids, 100 g
Purity: 99% min CAS No.: 7550-35-8 Molecular Formula: BrLi Formula Weight: 86.85 Physical Form: Crystalline Density: 3.464 Appearance: White Odour: Odourless Melting Point: 550° Boiling Point: 1265° Solubility: Very soluble in water and alcohol. Soluble in methanol, acetone, and glycol. Slightly soluble in pyridine Merck Reference: 14,5526 MDL No.: MFCD00011077
Pharmaceuticals, air-conditioning, humectant, drying agent, batteries, low temperature heat-exchange medium
- :7550-35-8
- :LiBr
- :MFCD00011077
- :86.84 g/mol
- :14,5526
- :1265 °C
- :550 °C
- :3,464
- :Stue temperatur
- :231-439-8
- Pakning:Glass bottle for solids
- Pakn:100 g