Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Borsyre ≥99.8%

Borsyre ≥99.8%

Glass bottle for solids, 1 kg
Katalog # 12680.A1
Sundhed
Leverandør:   Thermo Fisher Scientific
Borsyre ≥99.8%
Katalog # 12680.A1
CAS nummer:  10043-35-3
Borsyre ≥99.8%
Borsyre ≥99.8%

Glass bottle for solids, 1 kg

CAS nummer:  10043-35-3
Price
646,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

Purity: 99.8% CAS No.: 10043-35-3 Molecular Formula: BH3O3 Formula Weight: 61.83 Flash Point: None Physical Form: Granular Density: 1.435 Appearance: White Odour: Odourless pH: 3.8-4.8 Melting Point: ca 185° dec. Beilstein Reference: 1697939 Merck Reference: 14,1336 MDL No.: MFCD00011337

Synonymer:  Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
  • CAS nummer:10043-35-3
  • Formel:B(OH)₃
  • MDL nummer:MFCD00011337
  • Molecular Weight:61.83 g/mol
  • Merck Index:14,1336
  • Kogende Pt:185 °C (1013 hPa)
  • Smeltende Pt:∼ 185 °C (dec.)
  • Tæthed:1,435
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-139-2
  • NÅ:01-2119486683-25
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.