Zinc selenite
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 13597-46-1 Molecular Formula: ZnSeO3 Formula Weight: 192.33 Physical Form: Powder UN No.: 2630 MDL No.: MFCD00011297
- CAS Number:13597-46-1
- Formula:O₃SeZn
- MDL Number:MFCD00011297
- Molecular Weight:192.33 g/mol
- Storage Temperature:Stue temperatur
- EINECS:237-048-9
- UN:2630
- ADR:6.1,I
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:10 g



