Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ammoniumchlorid ≥99.5%

Ammoniumchlorid ≥99.5%

Plastic bottle for solids, 500 g
Katalog # 12361.36
Irriterende
Leverandør:   Thermo Fisher Scientific
Ammoniumchlorid ≥99.5%
Katalog # 12361.36
CAS nummer:  12125-02-9
Ammoniumchlorid ≥99.5%
Ammoniumchlorid ≥99.5%

Plastic bottle for solids, 500 g

CAS nummer:  12125-02-9
Price
551,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99.5% min CAS No.: 12125-02-9 Molecular Formula: NH4Cl Formula Weight: 53.49 Physical Form: Granular Density: ≈1.53 Refractive Index: 1.642 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 340° subl. Solubility: Soluble in water, methanol, ethanol. Insoluble in acetone, ether, ethyl acetate. Merck Reference: 14,509 MDL No.: MFCD00011420

As a flux in galvanizing steel sheet, in batteries, to clean soldering irons, analytical lab reagent, in washing powders, electroplating, lustering cotton

Synonymer:  Ammonium muriate, Salmiac
  • CAS nummer:12125-02-9
  • Formel:NH₄Cl
  • MDL nummer:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Merck Index:14,509
  • Smeltende Pt:340 °C (subl.)
  • Tæthed:1,53 g/cm³
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:235-186-4
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.