Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bariumnitrat 99+% ACS

Bariumnitrat 99+% ACS

Glass bottle for solids, 2 kg
Katalog # 12194.A3
Oxiderende
Irriterende
Leverandør:   Thermo Fisher Scientific
Bariumnitrat 99+% ACS
Katalog # 12194.A3
CAS nummer:  10022-31-8
Bariumnitrat 99+% ACS
Bariumnitrat 99+% ACS

Glass bottle for solids, 2 kg

CAS nummer:  10022-31-8
Price
2.190,00 kr
Log ind for tilgængelighed
1 * 2 kg
Product Details & Documents

Purity: 99+% CAS No.: 10022-31-8 Molecular Formula: Ba(NO3)2 Formula Weight: 261.35 Physical Form: Crystalline Density: 3.24 Melting Point: 592° dec. Solubility: Freely soluble in water. Slightly soluble in alcohol and acetone Merck Reference: 14,983 UN No.: 1446 MDL No.: MFCD00003442

  • CAS nummer:10022-31-8
  • Formel:Ba(NO₃)₂
  • MDL nummer:MFCD00003442
  • Molecular Weight:261.34 g/mol
  • Merck Index:14,983
  • Smeltende Pt:592 °C (dec.)
  • Tæthed:3,24
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-020-5
  • FN:1446
  • ADR:5.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.