Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Bariumnitrat 99+% ACS

Bariumnitrat 99+% ACS

Glass bottle for solids, 50 g
Katalog # 12194.18
Oxiderende
Irriterende
Leverandør:   Thermo Fisher Scientific
Bariumnitrat 99+% ACS
Katalog # 12194.18
CAS nummer:  10022-31-8
Bariumnitrat 99+% ACS
Bariumnitrat 99+% ACS

Glass bottle for solids, 50 g

CAS nummer:  10022-31-8
Price
353,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Purity: 99+% CAS No.: 10022-31-8 Molecular Formula: Ba(NO3)2 Formula Weight: 261.35 Physical Form: Crystalline Density: 3.24 Melting Point: 592° dec. Solubility: Freely soluble in water. Slightly soluble in alcohol and acetone Merck Reference: 14,983 UN No.: 1446 MDL No.: MFCD00003442

  • CAS nummer:10022-31-8
  • Formel:Ba(NO₃)₂
  • MDL nummer:MFCD00003442
  • Molecular Weight:261.34 g/mol
  • Merck Index:14,983
  • Smeltende Pt:592 °C (dec.)
  • Tæthed:3,24
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:233-020-5
  • FN:1446
  • ADR:5.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer