EDTA ≥99.4% ACS
Glass bottle for solids, 2,5 kg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.4+% CAS No.: 60-00-4 Molecular Formula: C10H16N2O8 Formula Weight: 292.24 Physical Form: Powder Appearance: White Melting Point: ca 245° dec. Beilstein Reference: 1716295 Merck Reference: 14,3517 MDL No.: MFCD00003541
Chelating agent
Synonyms: Kestranal® A, OmniPur® EDTA, Edetic acid, Ethylenediaminetetraacetic acid, Ethylenedinitrilotetra-acetic acid, 2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acid, (Ethylenedinitrilo)tetraacetic acid, Idranal® II
- CAS Number:60-00-4
- Formula:C₁₀H₁₆N₂O₈
- MDL Number:MFCD00003541
- Molecular Weight:292.25 g/mol
- Merck Index:14,3517
- Boiling Pt:614 °C (1013 hPa)
- Melting Pt:∼ 245 °C (dec.)
- Density:0,86 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Stue temperatur
- EINECS:200-449-4
- REACH:01-2119486399-18
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:2,5 kg



