Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Zinc phosphate hydrate, tech.

Zinc phosphate hydrate, tech.

500 g
Katalog # 11589.36
Leverandør:   Thermo Fisher Scientific
Zinc phosphate hydrate, tech.
Katalog # 11589.36
CAS nummer:  7779-90-0
Zinc phosphate hydrate, tech.
Zinc phosphate hydrate, tech.

500 g

CAS nummer:  7779-90-0
Price
963,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

CAS No.: 7779-90-0 Molecular Formula: Zn3(PO4)2.xH2O Formula Weight: 386.05(anhy) Physical Form: -325 Mesh Powder Solubility: Soluble in acetic acid, dilute mineral acids, ammonia. Insoluble in water UN No.: UN3077 MDL No.: MFCD00036282

  • CAS nummer:7779-90-0
  • Formel:O₈P₂Zn₃·xH₂O
  • MDL nummer:MFCD00036282
  • Molecular Weight:386.08 g/mol (anhy)
  • Merck Index:14,10151
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-944-3
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakn:
    500 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.