Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lithiumhexafluorphosphat ≥98%

Lithiumhexafluorphosphat ≥98%

Glass bottle for solids, 50 g
Katalog # 11529.18
Giftig
Ætsende
Leverandør:   Thermo Fisher Scientific
Lithiumhexafluorphosphat ≥98%
Katalog # 11529.18
CAS nummer:  21324-40-3
Lithiumhexafluorphosphat ≥98%
Lithiumhexafluorphosphat ≥98%

Glass bottle for solids, 50 g

CAS nummer:  21324-40-3
Price
3.830,00 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents

Purity: 98% CAS No.: 21324-40-3 Molecular Formula: LiPF6 Formula Weight: 151.90 Storage Temperature: 2°C to 8°C Physical Form: Powder Density: 1.5 Sensitivity: Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 200° dec. UN No.: 2923 MDL No.: MFCD00011096

Used as an electrolyte in Li-ion batteries.

Synonymer:  Lithium hexafluorophosphate(V)
  • CAS nummer:21324-40-3
  • Formel:F₆LiP
  • MDL nummer:MFCD00011096
  • Molecular Weight:151.91 g/mol
  • Smeltende Pt:200 °C (dec.)
  • Tæthed:1,5
  • Opbevaringstemperatur:Køleskab
  • EINECS:244-334-7
  • FN:2923
  • ADR:8,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer