Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lithiumhexafluorphosphat ≥98%

Lithiumhexafluorphosphat ≥98%

Glass bottle for solids, 10 g
Katalog # 11529.09
Giftig
Ætsende
Leverandør:   Thermo Fisher Scientific
Lithiumhexafluorphosphat ≥98%
Katalog # 11529.09
CAS nummer:  21324-40-3
Lithiumhexafluorphosphat ≥98%
Lithiumhexafluorphosphat ≥98%

Glass bottle for solids, 10 g

CAS nummer:  21324-40-3
Price
1.010,00 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents

Purity: 98% CAS No.: 21324-40-3 Molecular Formula: LiPF6 Formula Weight: 151.90 Storage Temperature: 2°C to 8°C Physical Form: Powder Density: 1.5 Sensitivity: Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 200° dec. UN No.: 2923 MDL No.: MFCD00011096

Used as an electrolyte in Li-ion batteries.

Synonymer:  Lithium hexafluorophosphate(V)
  • CAS nummer:21324-40-3
  • Formel:F₆LiP
  • MDL nummer:MFCD00011096
  • Molecular Weight:151.91 g/mol
  • Smeltende Pt:200 °C (dec.)
  • Tæthed:1,5
  • Opbevaringstemperatur:Køleskab
  • EINECS:244-334-7
  • FN:2923
  • ADR:8,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.