Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Lithiumhexafluorphosphat ≥98%

Lithiumhexafluorphosphat ≥98%

Glass bottle for solids, 1 g
Catalog # 11529.03
Toxic
Corrosive
Lithiumhexafluorphosphat ≥98%
Catalog # 11529.03
CAS Number:  21324-40-3
Lithiumhexafluorphosphat ≥98%
Lithiumhexafluorphosphat ≥98%

Glass bottle for solids, 1 g

CAS Number:  21324-40-3
Price
685,00 kr
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 98% CAS No.: 21324-40-3 Molecular Formula: LiPF6 Formula Weight: 151.90 Storage Temperature: 2°C to 8°C Physical Form: Powder Density: 1.5 Sensitivity: Moisture Sensitive Appearance: White Odour: Odourless Melting Point: 200° dec. UN No.: 2923 MDL No.: MFCD00011096

Used as an electrolyte in Li-ion batteries.

Synonyms:  Lithium hexafluorophosphate(V)
  • CAS Number:21324-40-3
  • Formula:F₆LiP
  • MDL Number:MFCD00011096
  • Molecular Weight:151.91 g/mol
  • Melting Pt:200 °C (dec.)
  • Density:1,5
  • Storage Temperature:Køleskab
  • EINECS:244-334-7
  • UN:2923
  • ADR:8,II
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifications