Calciumfluorid ≥99%
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: Transparent Crystals Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Odour: Odourless Melting Point: 1403° Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907
- CAS Number:7789-75-5
- Formula:CaF₂
- MDL Number:MFCD00010907
- Molecular Weight:78.07 g/mol
- Merck Index:14,1667
- Boiling Pt:∼2500 °C
- Melting Pt:1403 °C
- Density:3,18
- Storage Temperature:Stue temperatur
- EINECS:232-188-7
Documents
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:50 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



