Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Brom ≥99.8%, liquid

Brom ≥99.8%, liquid

5x1 kg, 5
Catalog # 00905.D9
Toxic
Corrosive
Environment
Brom ≥99.8%, liquid
Catalog # 00905.D9
CAS Number:  7726-95-6
Brom ≥99.8%, liquid
Brom ≥99.8%, liquid

5x1 kg, 5

CAS Number:  7726-95-6
Price
9.150,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents

Purity: 99.8% CAS No.: 7726-95-6 Physical Form: Liquid Odour: Unpleasant UN No.: 1744 MDL No.: MFCD00010896

  • CAS Number:7726-95-6
  • Formula:Br₂
  • MDL Number:MFCD00010896
  • Molecular Weight:159.81 g/mol
  • Merck Index:13,01378
  • Boiling Pt:58,7 °C (1013 hPa)
  • Melting Pt:–7,2 °C
  • Density:3,12 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:231-778-1
  • UN:1744
  • ADR:8,I
Specifications

Specifications

  • Pakn:
    5
  • Pk Info:
    5x1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.