Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Brom ≥99.8%, liquid

Brom ≥99.8%, liquid

Glass bottle for solids, 1 kg
Katalog # 00905.A1
Giftig
Ætsende
Miljø
Leverandør:   Thermo Fisher Scientific
Brom ≥99.8%, liquid
Katalog # 00905.A1
CAS nummer:  7726-95-6
Brom ≥99.8%, liquid
Brom ≥99.8%, liquid

Glass bottle for solids, 1 kg

CAS nummer:  7726-95-6
Price
2.860,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents

Purity: 99.8% CAS No.: 7726-95-6 Physical Form: Liquid Odour: Unpleasant UN No.: 1744 MDL No.: MFCD00010896

  • CAS nummer:7726-95-6
  • Formel:Br₂
  • MDL nummer:MFCD00010896
  • Molecular Weight:159.81 g/mol
  • Merck Index:13,01378
  • Kogende Pt:58,7 °C (1013 hPa)
  • Smeltende Pt:–7,2 °C
  • Tæthed:3,12 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-778-1
  • FN:1744
  • ADR:8,I
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.