Brom ≥99.8%, liquid
Glass bottle for solids, 250 g
Purity: 99.8% CAS No.: 7726-95-6 Physical Form: Liquid Odour: Unpleasant UN No.: 1744 MDL No.: MFCD00010896
- :7726-95-6
- :Br₂
- :MFCD00010896
- :159.81 g/mol
- :13,01378
- :58,7 °C (1013 hPa)
- :–7,2 °C
- :3,12 g/cm³ (20 °C)
- :Stue temperatur
- :231-778-1
- :1744
- :8,I
- Pakning:Glass bottle for solids
- Pakn:250 g



