Brom ≥99.8%, liquid
Glass bottle for solids, 25 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99.8% CAS No.: 7726-95-6 Physical Form: Liquid Odour: Unpleasant UN No.: 1744 MDL No.: MFCD00010896
- CAS nummer:7726-95-6
- Formel:Br₂
- MDL nummer:MFCD00010896
- Molecular Weight:159.81 g/mol
- Merck Index:13,01378
- Kogende Pt:58,7 °C (1013 hPa)
- Smeltende Pt:–7,2 °C
- Tæthed:3,12 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:231-778-1
- FN:1744
- ADR:8,I
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g



