Microscope adapters for micromanipulators
Nikon Eclipse® TS100
Catalog # 631-0217
Supplier: EPPENDORF
Jump to:
Product Details & Documents
Microscope adapters for InjectMan® NI 2 and TransferMan® NK 2. The universal stand is suitable for mounting manipulators on upright or stereo microscopes, independent of the microscope tripod used.
Specifications
- Beskrivelse:Nikon Eclipse® TS100