Orders LinkContactUsLinkComponent
Testkits, TOC til Spectroquant®, Supelco®
Testkits, TOC til Spectroquant®, Supelco®
TOC-kuvettetest*5, 50 - 800 mg/l TOC, Indicator, Prove, Nova 30/60, Multy, 25 Tests
Catalog # 1.14879.0001
Supplier:   Merck
Testkits, TOC til Spectroquant®, Supelco®
Catalog # 1.14879.0001
Supplier:  Merck
CAS Number:  
Testkits, TOC til Spectroquant®, Supelco®
Testkits, TOC til Spectroquant®, Supelco®
TOC-kuvettetest*5, 50 - 800 mg/l TOC, Indicator, Prove, Nova 30/60, Multy, 25 Tests
CAS Number:  
Price
2.790,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*5 TOC-skruelåg til nedbrydning påkrævet.
Documents
Specifications
Specifications
  • Kemisk metode:
    Indicator
  • Pakn:
    25 Tests
  • Beskrivelse:
    TOC-kuvettetest*<sup>5</sup>
  • Til enhed:
    Prove, Nova 30/60, Multy
  • Område:
    50 - 800 mg/l TOC
  • Cat. No.:
    1.14879.0001
  • M-Clarity quality level:
    MQ100
Specifications
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Availability
    Price
  • 1.73500.0001
    TOC, skruelåg til brug i Termoreactor
    Log in for availability
    1 * 6 Items - 1.050,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Koncentration
    Pakn
    Availability
    Price
  • 1.09017.0100
    TOC iflg. EN 1484 H3
    1000 (mg/l)
    100 mL
    Log in for availability
    1 * 100 mL - 1.610,00 kr