Pipettespidser, epT.I.P.S.® Standard
20 - 300 µl, -, 55 mm, Yellow, orange, Pose
These tips feature an ergonomically optimised cone geometry which provides an improved tip seal.
- Kvalitetsspidser
- Ergonomisk design
- Kegleform med smidig og bøjelig forseglingsområde, som giver en bedre forsegling ved pipetteskaftet
epT.I.P.S., in combination with adequate pipettes, are intended for dispensing liquids in the volume range of 0,1 µl to 10 ml.
In vivo applications (applications in or on the human body) are not allowed. epT.I.P.S. and the appropriate pipettes only must be used by correspondingly trained specialists.
The user must have read thoroughly the operating instructions of the pipette and the instructions for use of the epT.I.P.S. and must have made himself familiar with the functioning of the pipette with the pipette tips.
Packaging: Pakket i genlukkelige poser.
Documents
- Farvekode:orange
- Pakket:Pose
- Steril:-
- Volumen:20 - 300 µl
- Længde:55 mm
- Seq:07
- Spits farge:Yellow
- Cat. No.:613-3504
- Miljøcertificering:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Lav fremstillingspåvirkning