Large insulating dewar flasks, type 131 - 135
Large insulating dewar flasks, type 134
Katalog # KGWA1265
Leverandør: KGW
Gå til:
Product Details & Documents
Dewar flasks are made of glass and are reliable and economical with a protective casing out of blue-coated metal. These are used in laboratory techniques, medicinal techniques, biotechnology, storing of CO₂ and pressure-less space for coolants.
- With side grips
- Lid made of metal, blue coated with insulating disc
- Lid with over pressure valve
- Lockable lid and roller base
: Glass material according to ISO 3585 and Dewar flasks according to DIN 12492.
Specifikationer
- Beskrivelse:Large insulating dewar flasks, type 134
- Seq:02
- Cat. No.:KGWA1265
- Model:134
- Volumen:28 L
- Daglig fordampning:0,6 L
- Overall height:765 mm
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Insulating disc for type 131
Produktinformation
- Katalog #TilgængelighedPrice
- Insulating disc for type 133
Produktinformation
- Katalog #TilgængelighedPrice
- Insulating disc for type 135
Produktinformation
- Katalog #TilgængelighedPrice
- Linen-bag for type 132
Produktinformation
- Katalog #TilgængelighedPrice
- Linen-bag for type 135
Produktinformation
- Katalog #TilgængelighedPrice
- Insulating disc for type 132
Produktinformation
- Katalog #TilgængelighedPrice
- Linen-bag for type 131
Produktinformation
- Katalog #TilgængelighedPrice
- Linen-bag for type 133
Produktinformation
- Katalog #TilgængelighedPrice
- Linen-bag for type 134
Produktinformation
- Katalog #TilgængelighedPrice
- Insulating disc for type 134