Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Conductivity standards, Certipur® Supelco®

Conductivity standards, Certipur® Supelco®

KCl 0,001 M, 147 µS/cm, in 500 ml plastic bottle, Plastic bottle, 500 mL
Katalog # 1.01557.0500
Leverandør:   Merck
Conductivity standards, Certipur® Supelco®
Katalog # 1.01557.0500
Leverandør:  Merck
CAS nummer:  
Conductivity standards, Certipur® Supelco®
Conductivity standards, Certipur® Supelco®

KCl 0,001 M, 147 µS/cm, in 500 ml plastic bottle, Plastic bottle, 500 mL

CAS nummer:  
Price
1.870,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.

  • Traceable to PTB and NIST
  • Tested against primary reference standards

Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.

Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    500 mL
  • Beskrivelse:
    KCl 0,001 M, 147 µS/cm, in 500 ml plastic bottle
  • Cat. No.:
    1.01557.0500
  • M-Clarity quality level:
    MQ100

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.