pH and ORP electrodes
pH-glaselektrode med fri ende, flangelåg, Ag/AgCl-referencesystem, kuppelformet membran, H-glas, nulpunkt-pH = 7,0, 9501/85, 0 - 14, 0...+135 °C, Glas, 12×120 mm, 5 m fast kabel
Catalog # SCOI285114859
Supplier: SI Analytics
Jump to:
Product Details & Documents
pH electrodes with glass body.
Specifications
- Tilslutning:5 m fast kabel
- Ø×L:12×120 mm
- Model:9501/85
- Beskrivelse:pH-glaselektrode med fri ende, flangelåg, Ag/AgCl-referencesystem, kuppelformet membran, H-glas, nulpunkt-pH = 7,0
- Materiale:Glas
- Seq:02
- Temperatur:0...+135 °C
- pH:0 - 14