Conductivity standards, Certipur® Supelco®
KCl 0,1 M, 12 800 µS/cm, sachets of 30 ml, Sachet, 30
: Merck
These conductivity sachets are convenient, easy to use and ensure accurate and reliable results both in the field or in the laboratory. The 30 ml sachets always provide a fresh solution preventing contamination by microorganisms, CO₂ or other substances. A conductivity sensor can be simply inserted into the sachet.
- Traceable to PTB and NIST
- Tested against primary reference standards
Certipur conductivity standards are also available in 500 ml packs for the user with higher volume requirements.
Documents
- Pakning:Sachet
- Pakn:30
- Beskrivelse:KCl 0,1 M, 12 800 µS/cm, sachets of 30 ml
- Cat. No.:1.01554.0001
- M-Clarity quality level:MQ100
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



