Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Nitrite is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications. Nitrites must be closely monitored in drinking and groundwater as well as in process, waste, or aquarium water. In farm produce and fruit juices, high nitrate concentrations affect quality and keeping properties and can constitute serious health risks. Nitrites are also used in additives for food products and serve as corrosion inhibitors in engine cooling systems.
Documents
Specifikationer
Specifikationer
- Kemisk metode:Aromatic amine
- Pakn:50 Tests
- Beskrivelse:Nitrattest
- Område:30 - 1000 mg/l NO₂
- Cat. No.:1.16732.0001
- M-Clarity quality level:MQ100
Specifikationer
Complementary Products
Produktinformation
- Katalog #BeskrivelseModelTilgængelighedPrice
- 1.17246.0001Reflectometer for test strips onlyRQflex 20Log ind for tilgængelighed1 * 1 Items - 10.580,00 kr