Orders LinkContactUsLinkComponent
Reflectoquant® teststrimler, Supelco®
Reflectoquant® teststrimler, Supelco®
Nitrattest, 30 - 1000 mg/l NO₂, Aromatic amine, 50 Tests
Katalog # 1.16732.0001
Leverandør:   Merck
Reflectoquant® teststrimler, Supelco®
Katalog # 1.16732.0001
Leverandør:  Merck
CAS nummer:  
Reflectoquant® teststrimler, Supelco®
Reflectoquant® teststrimler, Supelco®
Nitrattest, 30 - 1000 mg/l NO₂, Aromatic amine, 50 Tests
CAS nummer:  
Price
844,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly
Nitrite is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications. Nitrites must be closely monitored in drinking and groundwater as well as in process, waste, or aquarium water. In farm produce and fruit juices, high nitrate concentrations affect quality and keeping properties and can constitute serious health risks. Nitrites are also used in additives for food products and serve as corrosion inhibitors in engine cooling systems.
Specifikationer
Specifikationer
  • Kemisk metode:
    Aromatic amine
  • Pakn:
    50 Tests
  • Beskrivelse:
    Nitrattest
  • Område:
    30 - 1000 mg/l NO₂
  • Cat. No.:
    1.16732.0001
  • M-Clarity quality level:
    MQ100
Specifikationer
Complementary Products
Produktinformation
  • Katalog #
    Beskrivelse
    Model
    Tilgængelighed
    Price
  • 1.17246.0001
    Reflectometer for test strips only
    RQflex 20
    Log ind for tilgængelighed
    1 * 1 Items - 10.580,00 kr