Buffer solutions, according to DIN 19266, Fluka™
7,413, —, Fosfat, 25 °C, Plastic bottle, 1×1 L, 1 L
Katalog # 33597-1L
Leverandør: Honeywell Chemicals
Buffer solutions, according to DIN 19266, Fluka™
Katalog # 33597-1L
Leverandør: Honeywell Chemicals
CAS nummer:
Gå til:
Product Details & Documents
Buffers with fungicide (except pH 10,012)
: According to DIN 19266.
Documents
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakning:Plastic bottle
- pH buffer type:Fosfat
- Pakn:1 L
- Referencetemperatur:25 °C
- pH nøjagtighed:—
- Pk Info:1×1 L
- pH-værdi:7,413
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



