Serological pipettes
2 ml, 0,01 ml, +, grøn, 25/pakke, 1000/kasse
Katalog # SARS86.1252.025
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
PS, graduated, sterile.
- Plugged
- Easy identification by colour code
- Larger pipetting volume due to negative graduation
- Optimised mouth piece for universal fit in most common pipettors
- Easy to open, static-free packaging
Documents
Specifikationer
- Farvekode:grøn
- Pakket:25/pakke, 1000/kasse
- Steril:+
- Kapacitet:2 ml
- Inddeling:0,01 ml