Orders LinkContactUsLinkComponent
Double spatulas, micro, with one spoon end
Double spatulas, micro, with one spoon end
150 mm, 5 mm
Katalog # RSGA395.150
Leverandør:   RSG
Double spatulas, micro, with one spoon end
Katalog # RSGA395.150
Leverandør:  RSG
Double spatulas, micro, with one spoon end
Double spatulas, micro, with one spoon end
150 mm, 5 mm
Price
237,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
Micro double spatulas with one spoon shaped end and one spatula end. Made of stainless steel.
Specifikationer
  • Længde:
    150 mm
  • Seq:
    1
  • Bladbredde:
    5 mm