Laboratorieknive
Broad blade, 150 mm
233-0149
: RSG
These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
- Anti-magnetic and lightweight
- Dishwasher-proof
- Long-life sharpness
- Model:Broad blade
- Seq:04
- Klinge:150 mm