Laboratorieknive
Broad blade, 90 mm
Katalog # 233-0147
Leverandør: RSG
Gå til:
Product Details & Documents
These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
- Anti-magnetic and lightweight
- Dishwasher-proof
- Long-life sharpness
Specifikationer
- Model:Broad blade
- Seq:02
- Klinge:90 mm