Laboratorieknive
Thin blade, 90 mm
Katalog # 233-0146
Leverandør: RSG
Gå til:
Product Details & Documents
These ceramic laboratory knives have extremely hard blades which resist chemicals, fractures and corrosion.
- Anti-magnetic and lightweight
- Dishwasher-proof
- Long-life sharpness
Specifikationer
- Model:Thin blade
- Seq:01
- Klinge:90 mm