Orders LinkContactUsLinkComponent
Dishes, crystallising
Dishes, crystallising
5000 ml, 160 mm, 200 mm
Katalog # RSGA072.005
Leverandør:   RSG
Dishes, crystallising
Katalog # RSGA072.005
Leverandør:  RSG
Dishes, crystallising
Dishes, crystallising
5000 ml, 160 mm, 200 mm
Price
3.530,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Made of stainless steel 18/10.
  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Specifikationer
  • Udv.Ø:
    200 mm
  • Kapacitet:
    5000 ml
  • Højde:
    160 mm
  • Seq:
    03