Safety Barrels
Safety barrel with screw cap 1½", 10 L
Catalog # 119-0115
Supplier: ROETZMEIER SICHERH BEHAELTER
Jump to:
Product Details & Documents
These safety barrels are suitable for safe and comfortable handling and storage of larger amounts of flammable liquid.
- In-plant use of flammable liquids such as solvents
- Explosion-proof due to flame traps (removable) and pressure control valves
- Taps and metering devices are self-closing
- High quality, chemically resistant stainless steel (thickness 0,8 to 1,5 mm depending on capacity) with PTFE gaskets
- Models with self-closing tap are equipped with separate ventilation which maximises the flow rate
- PFA content level indicator incl. a scale for volumes 10, 25 and 50 litres
Caution: * Collection barrel
Specifications
- Kapacitet:10 L
- Beskrivelse:Safety barrel with screw cap 1½"
- Seq:03
Accessories
Product Information
- Catalog #BeskrivelseDiameter×HTilAvailabilityPrice
- 119-0161Rotable spout 1½“28×175 mmAll Rötzmeier safety canisters and barrels (10 and 25 l)Log in for availability1 * 1 Items - 665,00 kr
Product Information
- Catalog #BeskrivelseDiameter×HTilAvailabilityPrice
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-0137Screw cap 2" TPI with pressure control valve and ventilation119-0127, -0128Log in for availability1 * 1 Items - 608,00 kr
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-0136Screw cap 2" TPI with pressure control valve119-0126Log in for availability1 * 1 Items - 600,00 kr
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-2400Self-closing tap ¾"119-0119, -0124, -0127, -0120, -0125, -0128Log in for availability1 * 1 Items - 1.190,00 kr
Product Information
- Catalog #BeskrivelseKapacitetAvailabilityPrice
- 119-0143Soaking tank for controlled moistening, soaking or bathing of particels, cloths, flannels, paper, etc., e.g. for cleaning with solvents4 lLog in for availability1 * 1 Items - 1.720,00 kr
Product Information
- Catalog #BeskrivelseKapacitetAvailabilityPrice
- 119-0154Washing tank for safe cleaning of particels and tools with inflammable liquids4 lLog in for availability1 * 1 Items - 468,00 kr
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-2391Self-closing metering device 1½“ for safety canisters and barrels119-0172, -0116, -2045, -2046, -2370, -2371, -0115Log in for availability1 * 1 Items - 714,00 kr
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-0169Flame trapAll Rötzmeier safety canisters (5, 10 and 20 l) and safety barrels (10 and 25 l)Log in for availability1 * 1 Items - 206,00 kr
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-0159Screw cap 1½" with pressure control valve and ventilation119-0119, -0124, -0120, -0125Log in for availability1 * 1 Items - 526,00 kr
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-0129Screw cap 1½" with pressure control valve119-0172, -0116, -2380, -2045, -2046, -0121, -2180, -2181, -2182, -2160, -0111, -0115, -0123Log in for availability1 * 1 Items - 517,00 kr
Product Information
- Catalog #BeskrivelseTilAvailabilityPrice
- 119-0163Self-closing metering device 1½" for safety canisters and barrels with sep. ventilation119-2160, -0111, -2380, -0122Log in for availability1 * 1 Items - 723,00 kr