Orders LinkContactUsLinkComponent
Safety Barrels
Safety Barrels
Safety barrel with screw cap 1½", 10 L
Catalog # 119-0115
Safety Barrels
Catalog # 119-0115
Safety Barrels
Safety Barrels
Safety barrel with screw cap 1½", 10 L
Price
2.260,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These safety barrels are suitable for safe and comfortable handling and storage of larger amounts of flammable liquid.
  • In-plant use of flammable liquids such as solvents
  • Explosion-proof due to flame traps (removable) and pressure control valves
  • Taps and metering devices are self-closing
  • High quality, chemically resistant stainless steel (thickness 0,8 to 1,5 mm depending on capacity) with PTFE gaskets
  • Models with self-closing tap are equipped with separate ventilation which maximises the flow rate
  • PFA content level indicator incl. a scale for volumes 10, 25 and 50 litres
Caution: * Collection barrel
Documents
Specifications
  • Kapacitet:
    10 L
  • Beskrivelse:
    Safety barrel with screw cap 1½"
  • Seq:
    03
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Diameter×H
    Til
    Availability
    Price
  • 119-0161
    Rotable spout 1½“
    28×175 mm
    All Rötzmeier safety canisters and barrels (10 and 25 l)
    Log in for availability
    1 * 1 Items - 665,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Diameter×H
    Til
    Availability
    Price
  • 119-0162
    Spout 2“
    40×180 mm
    119-0117, -0118
    Log in for availability
    1 * 1 Items - 444,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0137
    Screw cap 2" TPI with pressure control valve and ventilation
    119-0127, -0128
    Log in for availability
    1 * 1 Items - 608,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0136
    Screw cap 2" TPI with pressure control valve
    119-0126
    Log in for availability
    1 * 1 Items - 600,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-2400
    Self-closing tap ¾"
    119-0119, -0124, -0127, -0120, -0125, -0128
    Log in for availability
    1 * 1 Items - 1.190,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Kapacitet
    Availability
    Price
  • 119-0143
    Soaking tank for controlled moistening, soaking or bathing of particels, cloths, flannels, paper, etc., e.g. for cleaning with solvents
    4 l
    Log in for availability
    1 * 1 Items - 1.720,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Kapacitet
    Availability
    Price
  • 119-0154
    Washing tank for safe cleaning of particels and tools with inflammable liquids
    4 l
    Log in for availability
    1 * 1 Items - 468,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-2391
    Self-closing metering device 1½“ for safety canisters and barrels
    119-0172, -0116, -2045, -2046, -2370, -2371, -0115
    Log in for availability
    1 * 1 Items - 714,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0168
    Flame trap
    119-0126, -0127, -0128
    Log in for availability
    1 * 1 Items - 235,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0169
    Flame trap
    All Rötzmeier safety canisters (5, 10 and 20 l) and safety barrels (10 and 25 l)
    Log in for availability
    1 * 1 Items - 206,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0210
    Screw cap 2" Tri-Sure
    119-0117, -0118
    Log in for availability
    1 * 1 Items - 90,30 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0159
    Screw cap 1½" with pressure control valve and ventilation
    119-0119, -0124, -0120, -0125
    Log in for availability
    1 * 1 Items - 526,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0129
    Screw cap 1½" with pressure control valve
    119-0172, -0116, -2380, -2045, -2046, -0121, -2180, -2181, -2182, -2160, -0111, -0115, -0123
    Log in for availability
    1 * 1 Items - 517,00 kr
Product Information
  • Catalog #
    Beskrivelse
    Til
    Availability
    Price
  • 119-0163
    Self-closing metering device 1½" for safety canisters and barrels with sep. ventilation
    119-2160, -0111, -2380, -0122
    Log in for availability
    1 * 1 Items - 723,00 kr