Deflagrating spoon
Cap (90 mm) and cork only, for deflagrating spoon, -
Catalog # RLAAZ4804
Supplier: R L ENTERPRISES
Jump to:
Product Details & Documents
Mild steel. Deflagrating spoon, welded to a handle.
With cork and aluminium convex cap, 90 mm diameter to give safety closure.
Specifications
- Beskrivelse:Cap (90 mm) and cork only, for deflagrating spoon
- Længde:-
- Seq:03