Orders LinkContactUsLinkComponent
Deflagrating spoon
Deflagrating spoon
Cap (90 mm) and cork only, for deflagrating spoon, -
Catalog # RLAAZ4804
Supplier:   R L ENTERPRISES
Deflagrating spoon
Catalog # RLAAZ4804
Supplier:  R L ENTERPRISES
Deflagrating spoon
Cap (90 mm) and cork only, for deflagrating spoon, -
Price
58,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Mild steel. Deflagrating spoon, welded to a handle.
With cork and aluminium convex cap, 90 mm diameter to give safety closure.
Specifications
  • Beskrivelse:
    Cap (90 mm) and cork only, for deflagrating spoon
  • Længde:
    -
  • Seq:
    03