Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Deflagrating spoon

Deflagrating spoon only, without cork and cap, 400 mm
  RLAAZ4704
Deflagrating spoon
  RLAAZ4704
Deflagrating spoon

Deflagrating spoon only, without cork and cap, 400 mm

 
53,70 kr
   
1 * 1 Items
 

Mild steel. Deflagrating spoon, welded to a handle.

With cork and aluminium convex cap, 90 mm diameter to give safety closure.

 
  • Beskrivelse:
    Deflagrating spoon only, without cork and cap
  • Længde:
    400 mm
  • Seq:
    02